siRNA Details
| Gene Symbol: ZCRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033114.3 | NP_149105.3 | 4 | 423 | ||
| XM_017020124.1 | XP_016875613.1 | 3 | 314 | |||
| GenBank mRNA | AB062361.1 | - | 4 | 423 | ||
| AK308244.1 | - | 4 | 293 | |||
| BC005099.1 | - | 4 | 373 | |||
| BC010177.2 | - | 4 | 388 | |||
| BC022543.1 | - | 4 | 398 | 4797962 | ||

ZCRB1
Genomic Map