siRNA Details
| Gene Symbol: PURB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033224.4 | NP_150093.1 | 1 | 905 | ||
| GenBank mRNA | AY039216.1 | - | 1 | 881 | ||
| BC101735.1 | - | 1 | 887 | 8069241 | ||
| BC101737.1 | - | 1 | 887 | 8069243 | ||
| BU177276.1 | - | 1 | 305 | |||

PURB
Genomic Map