siRNA Details
| Gene Symbol: RASL10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033315.3 | NP_201572.1 | 4 | 1822 | ||
| XM_017025300.1 | XP_016880789.1 | 5 | 2716 | |||
| XM_017025301.1 | XP_016880790.1 | 3 | 2456 | |||
| GenBank mRNA | AK122652.1 | - | 4 | 1820 | ||
| BC041133.1 | - | 4 | 1742 | 5762715 | ||
| DA250459.1 | - | 1 | 419 | |||

RASL10B
Genomic Map