siRNA Details
| Gene Symbol: FATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033085.2 | NP_149076.1 | 5 | 763 | ||
| GenBank mRNA | AF249872.1 | - | 5 | 763 | ||
| AY101182.1 | - | 5 | 791 | |||
| BC022064.2 | - | 5 | 764 | 4826440 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FATE1| Gene Symbol: FATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033085.2 | NP_149076.1 | 5 | 763 | ||
| GenBank mRNA | AF249872.1 | - | 5 | 763 | ||
| AY101182.1 | - | 5 | 791 | |||
| BC022064.2 | - | 5 | 764 | 4826440 | ||