siRNA Details
| Gene Symbol: ZCRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033114.3 | NP_149105.3 | 5 | 510 | ||
| XM_017020124.1 | XP_016875613.1 | 4 | 401 | |||
| GenBank mRNA | AB062361.1 | - | 5 | 510 | ||
| AK308244.1 | - | 5 | 380 | |||
| BC005099.1 | - | 5 | 460 | |||
| BC010177.2 | - | 5 | 475 | |||
| BC022543.1 | - | 5 | 485 | 4797962 | ||
| BI666919.1 | - | 1 | 49 | |||

ZCRB1
Genomic Map