siRNA Details
| Gene Symbol: CCDC65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286957.1 | NP_001273886.1 | 3, 4 | 665 | ||
| NM_033124.4 | NP_149115.2 | 3, 4 | 705 | |||
| GenBank mRNA | AF382188.1 | - | 3, 4 | 670 | ||
| AK093051.1 | - | 3, 4 | 678 | |||
| AK302082.1 | - | 3, 4 | 642 | |||
| AK314623.1 | - | 3, 4 | 650 | |||
| BC039317.1 | - | 3, 4 | 644 | 5271866 | ||

CCDC65
Genomic Map