siRNA Details
| Gene Symbol: CCDC102A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033212.3 | NP_149989.2 | 9 | 2398 | ||
| XM_011523469.2 | XP_011521771.1 | 9 | 3678 | |||
| GenBank mRNA | BC009941.2 | - | 9 | 2398 | 4100726 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC102A| Gene Symbol: CCDC102A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033212.3 | NP_149989.2 | 9 | 2398 | ||
| XM_011523469.2 | XP_011521771.1 | 9 | 3678 | |||
| GenBank mRNA | BC009941.2 | - | 9 | 2398 | 4100726 | |