siRNA Details
| Gene Symbol: MRPS36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033281.5 | NP_150597.1 | 3 | 264 | ||
| GenBank mRNA | AF086034.1 | - | 2 | 95 | ||
| AF271777.1 | - | 368 | ||||
| BC015966.1 | - | 3 | 254 | 4068931 | ||
| BC017900.1 | - | 3 | 194 | 4251462 | ||
| BX951861.1 | - | 3 | 270 | |||

MRPS36
Genomic Map