siRNA Details
| Gene Symbol: ZNF160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102603.1 | NP_001096073.1 | 7 | 3519 | ||
| NM_001322128.1 | NP_001309057.1 | 7 | 3527 | |||
| NM_001322129.1 | NP_001309058.1 | 6 | 3622 | |||
| NM_001322130.1 | NP_001309059.1 | 7 | 3751 | |||
| NM_001322131.1 | NP_001309060.1 | 6 | 3626 | |||
| NM_001322132.1 | NP_001309061.1 | 7 | 3515 | |||
| NM_001322133.1 | NP_001309062.1 | 7 | 3539 | |||
| NM_001322134.1 | NP_001309063.1 | 7 | 3543 | |||
| NM_001322135.1 | NP_001309064.1 | 6 | 3398 | |||
| NM_001322136.1 | NP_001309065.1 | 5 | 3255 | |||
| NM_001322137.1 | NP_001309066.1 | 8 | 3575 | |||
| NM_001322138.1 | NP_001309067.1 | 7 | 3458 | |||
| NM_001322139.1 | NP_001309068.1 | 9 | 3672 | |||
| NM_033288.3 | NP_150630.2 | 6 | 3402 | |||
| NM_198893.2 | NP_942596.1 | 7 | 3487 | |||
| XM_017027445.1 | XP_016882934.1 | 4 | 3751 | |||
| XM_017027446.1 | XP_016882935.1 | 6 | 3712 | |||
| GenBank mRNA | AB046831.1 | - | 3 | 3872 | ||
| AK024442.1 | - | 1 | 2766 | |||
| BC094880.1 | - | 7 | 3404 | 3066532 | ||
| CR749613.1 | - | 1 | 2217 | |||
| X77744.1 | - | 1 | 1405 | |||

ZNF160
Genomic Map