siRNA Details
| Gene Symbol: HELZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037335.2 | NP_001032412.2 | 7 | 3383 | ||
| NM_033405.3 | NP_208384.3 | 1 | 888 | |||
| GenBank mRNA | AB051556.2 | - | 1 | 888 | ||
| AB201715.1 | - | 6 | 2561 | |||
| AF517673.1 | - | 1 | 1208 | |||
| BC150283.1 | - | 1 | 888 | 9007272 | ||

HELZ2
Genomic Map