siRNA Details
| Gene Symbol: CTTNBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033427.2 | NP_219499.1 | 1, 2 | 175 | ||
| XM_005250635.3 | XP_005250692.1 | 1, 2 | 107 | |||
| XM_011516616.2 | XP_011514918.1 | 1, 2 | 107 | |||
| XM_011516618.2 | XP_011514920.1 | 1, 2 | 107 | |||
| XM_017012706.1 | XP_016868195.1 | 1, 2 | 107 | |||
| XM_017012707.1 | XP_016868196.1 | 1, 2 | 107 | |||
| GenBank mRNA | AB051545.1 | - | 1, 2 | 81 | ||
| AF377960.1 | - | 1, 2 | 175 | |||
| AK091548.1 | - | 1, 2 | 109 | |||
| AK308081.1 | - | 1, 2 | 107 | |||
| BC106000.1 | - | 1, 2 | 142 | 30528261 | ||

CTTNBP2
Genomic Map