siRNA Details
| Gene Symbol: SLC38A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033518.3 | NP_277053.2 | 9 | 693 | ||
| XM_005272694.3 | XP_005272751.2 | 8 | 683 | |||
| XM_005272695.4 | XP_005272752.3 | 8 | 685 | |||
| XM_005272697.2 | XP_005272754.2 | 8 | 636 | |||
| XM_005272698.4 | XP_005272755.2 | 8 | 574 | |||
| XM_006724569.3 | XP_006724632.1 | 9 | 693 | |||
| XM_017029960.1 | XP_016885449.1 | 8 | 605 | |||
| XM_017029961.1 | XP_016885450.1 | 8 | 577 | |||
| GenBank mRNA | AF276889.1 | - | 8 | 1358 | ||
| AF318358.1 | - | 5 | 804 | |||
| AK094788.1 | - | 8 | 612 | |||
| BC019246.2 | - | 8 | 543 | 3506044 | ||
| BC027721.2 | - | 4 | 234 | |||

SLC38A5
Genomic Map