siRNA Details
| Gene Symbol: INTS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033547.3 | NP_291025.3 | 2 | 246 | ||
| XM_011545352.2 | XP_011543654.1 | 2 | 245 | |||
| XM_011545353.2 | XP_011543655.1 | 2 | 424 | |||
| XM_017018560.1 | XP_016874049.1 | 2 | 279 | |||
| GenBank mRNA | AK000455.1 | - | 2 | 228 | ||
| AK290866.1 | - | 2 | 242 | |||
| AL834171.1 | - | 2 | 189 | |||
| BC009995.2 | - | 2 | 241 | 4129693 | ||
| BC015664.2 | - | 2 | 246 | 4554197 | ||
| BC018672.1 | - | 2 | 228 | |||

INTS4
Genomic Map