siRNA Details
| Gene Symbol: ZNF764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172679.1 | NP_001166150.1 | 3 | 1657 | ||
| NM_033410.3 | NP_219363.2 | 3 | 1660 | |||
| GenBank mRNA | AK093992.1 | - | 3 | 1622 | ||
| BC000016.2 | - | 1 | 893 | |||
| BC008821.2 | - | 3 | 1602 | 4130947 | ||

ZNF764
Genomic Map