siRNA Details
| Gene Symbol: SNX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042632.2 | NP_001036097.1 | 4 | 2247 | ||
| NM_001042633.2 | NP_001036098.1 | 5 | 2265 | |||
| NM_033421.3 | NP_219489.1 | 4 | 2254 | |||
| NM_152897.2 | NP_690857.1 | 5 | 1002 | |||
| XM_005260608.4 | XP_005260665.1 | 2 | 2329 | |||
| XM_011529095.2 | XP_011527397.1 | 2 | 2233 | |||
| GenBank mRNA | AF258570.1 | - | 1 | 1915 | ||
| AK091359.1 | - | 3 | 2269 | |||
| AK095851.1 | - | 3 | 2125 | |||
| BC019823.1 | - | 5 | 975 | 4453502 | ||

SNX21
Genomic Map