siRNA Details
| Gene Symbol: SEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052838.4 | NP_443070.5 | 8 | 771 | ||
| XM_017022991.1 | XP_016878480.1 | 8 | 751 | |||
| GenBank mRNA | AF085235.1 | - | 7 | 602 | ||
| AL833004.1 | - | 2 | 1537 | |||
| AY034176.1 | - | 7 | 605 | |||
| BC012161.1 | - | 6 | 593 | 4576220 | ||
| BQ072902.1 | - | 8 | 785 | |||

SEPT1
Genomic Map