siRNA Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172831.1 | NP_001166302.1 | 7 | 753 | ||
| NM_001172832.1 | NP_001166303.1 | 5 | 656 | |||
| NM_052860.2 | NP_443092.1 | 6 | 698 | |||
| XM_011537702.2 | XP_011536004.1 | 7 | 853 | |||
| GenBank mRNA | AF395541.1 | - | 6 | 544 | ||
| AK095642.1 | - | 2 | 1445 | |||
| AK096465.1 | - | 6 | 695 | |||
| AK300528.1 | - | 7 | 674 | |||
| BC117248.1 | - | 6 | 447 | 40125799 | ||
| CR933727.1 | - | 1 | 165 | |||
| DQ908918.1 | - | 7 | 559 | |||

ZNF300
Genomic Map