siRNA Details
| Gene Symbol: MBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052897.3 | NP_443129.3 | 12 | 3136 | ||
| XM_005268617.1 | XP_005268674.1 | 12 | 3192 | |||
| XM_005268618.1 | XP_005268675.1 | 12 | 3288 | |||
| XM_006719217.1 | XP_006719280.1 | 12 | 3037 | |||
| XM_006719218.1 | XP_006719281.1 | 12 | 3025 | |||
| XM_011537851.2 | XP_011536153.1 | 12 | 4025 | |||
| GenBank mRNA | AB067474.1 | - | 9 | 2805 | ||
| AK056399.1 | - | 12 | 3170 | |||
| AK093078.1 | - | 7 | 2034 | |||
| AL834230.1 | - | 6 | 1415 | |||
| BC065530.1 | - | 12 | 3136 | 6152748 | ||
| CR627478.1 | - | 5 | 715 | |||

MBD6
Genomic Map