siRNA Details
| Gene Symbol: RGS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102450.2 | NP_001095920.1 | 7 | 790 | ||
| NM_033345.3 | NP_203131.1 | 6 | 661 | |||
| XM_005245555.3 | XP_005245612.1 | 7 | 804 | |||
| XM_011510089.2 | XP_011508391.1 | 7 | 1701 | |||
| XM_011510090.2 | XP_011508392.1 | 10 | 1531 | |||
| XM_011510091.2 | XP_011508393.1 | 6 | 678 | |||
| XM_017002631.1 | XP_016858120.1 | 10 | 1151 | |||
| XM_017002632.1 | XP_016858121.1 | 8 | 982 | |||
| XM_017002633.1 | XP_016858122.1 | 7 | 1180 | |||
| XM_017002634.1 | XP_016858123.1 | 7 | 859 | |||
| XM_017002635.1 | XP_016858124.1 | 8 | 1306 | |||
| XM_017002636.1 | XP_016858125.1 | 6 | 788 | |||
| XM_017002637.1 | XP_016858126.1 | 7 | 986 | |||
| GenBank mRNA | AF300649.1 | - | 5 | 499 | ||
| AK294660.1 | - | 7 | 757 | |||
| AK296514.1 | - | 7 | 750 | |||
| BC069677.1 | - | 7 | 739 | 7262579 | ||
| BC069718.1 | - | 6 | 611 | 7262591 | ||
| BC103865.1 | - | 7 | 766 | 40003386 | ||
| BC103866.1 | - | 7 | 766 | 40003387 | ||
| BC103867.1 | - | 7 | 766 | 40003388 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026380, BC065056, AK142301, AK044337, BC094577.

RGS8
Genomic Map