siRNA Details
Gene Symbol: ZNF747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305018.1 | NP_001291947.1 | 3 | 2371 | ||
NM_001305019.1 | NP_001291948.1 | 3 | 2368 | |||
NM_001305020.1 | NP_001291949.1 | 2 | 2508 | |||
NM_023931.3 | NP_076420.1 | 2 | 2511 | |||
GenBank mRNA | AA316824.1 | - | 1 | 246 | ||
BM461939.1 | - | 1 | 68 |
Gene Symbol: ZNF764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172679.1 | NP_001166150.1 | 3 | 2321 | ||
NM_033410.3 | NP_219363.2 | 3 | 2324 | |||
GenBank mRNA | AK093992.1 | - | 3 | 2286 | ||
BC000016.2 | - | 1 | 1557 | |||
BC008821.2 | - | 3 | 2266 | 4130947 |
