siRNA Details
| Gene Symbol: ARMC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199196.1 | NP_001186125.1 | 9 | 2458 | ||
| NM_033415.3 | NP_219483.1 | 8 | 2350 | |||
| XM_005260157.3 | XP_005260214.1 | 8 | 2401 | |||
| XM_017027484.1 | XP_016882973.1 | 8 | 2348 | |||
| XM_017027485.1 | XP_016882974.1 | 10 | 2582 | |||
| GenBank mRNA | BC003700.1 | - | 4 | 1096 | ||
| BC056161.1 | - | 10 | 2080 | |||
| BC067125.1 | - | 9 | 2227 | 6161112 | ||
| BX648486.1 | - | 8 | 2245 | |||
| CR749678.1 | - | 5 | 1689 | |||

ARMC6
Genomic Map