siRNA Details
| Gene Symbol: SNX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042632.2 | NP_001036097.1 | 4 | 2930 | ||
| NM_001042633.2 | NP_001036098.1 | 5 | 2948 | |||
| NM_033421.3 | NP_219489.1 | 4 | 2937 | |||
| NM_152897.2 | NP_690857.1 | 5 | 1685 | |||
| GenBank mRNA | AK095851.1 | - | 3 | 2808 | ||

SNX21
Genomic Map