siRNA Details
| Gene Symbol: INTS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033547.3 | NP_291025.3 | 6 | 712 | ||
| XM_011545352.2 | XP_011543654.1 | 7 | 881 | |||
| XM_011545353.2 | XP_011543655.1 | 6 | 890 | |||
| XM_017018560.1 | XP_016874049.1 | 6 | 745 | |||
| GenBank mRNA | AK290866.1 | - | 6 | 708 | ||
| AL834171.1 | - | 6 | 655 | |||
| BC008013.1 | - | 2 | 165 | |||
| BC009995.2 | - | 6 | 707 | 4129693 | ||
| BC015664.2 | - | 5 | 605 | 4554197 | ||

INTS4
Genomic Map