siRNA Details
| Gene Symbol: FBXO21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015002.2 | NP_055817.1 | 12 | 2942 | ||
| NM_033624.2 | NP_296373.1 | 12 | 2963 | |||
| XM_017019037.1 | XP_016874526.1 | 11 | 2788 | |||
| XM_017019038.1 | XP_016874527.1 | 11 | 2767 | |||
| XM_017019039.1 | XP_016874528.1 | 11 | 2558 | |||
| GenBank mRNA | AB020682.1 | - | 12 | 2904 | ||

FBXO21
Genomic Map