siRNA Details
| Gene Symbol: MGME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310338.1 | NP_001297267.1 | 5, 6 | 1227 | ||
| NM_052865.3 | NP_443097.1 | 4, 5 | 1182 | |||
| XM_005260867.2 | XP_005260924.1 | 5, 6 | 1449 | |||
| XM_011529394.1 | XP_011527696.1 | 5, 6 | 1064 | |||
| XM_011529395.1 | XP_011527697.1 | 4, 5 | 942 | |||
| XM_017028127.1 | XP_016883616.1 | 4, 5 | 1404 | |||
| XM_017028128.1 | XP_016883617.1 | 4, 5 | 1278 | |||
| GenBank mRNA | AF306684.1 | - | 4, 5 | 991 | ||
| AF306685.1 | - | 4, 5 | 991 | |||
| AK027503.1 | - | 4, 5 | 986 | |||
| AK309563.1 | - | 2, 3 | 525 | |||
| AK309995.1 | - | 2, 3 | 453 | |||
| AK310282.1 | - | 3, 4 | 498 | |||
| AK315532.1 | - | 4, 5 | 1018 | |||
| BC016869.1 | - | 4, 5 | 954 | 3850853 | ||

MGME1
Genomic Map