siRNA Details
| Gene Symbol: C8orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052958.2 | NP_443190.2 | 8 | 1430 | ||
| XM_011517445.2 | XP_011515747.1 | 8 | 1402 | |||
| XM_011517446.2 | XP_011515748.1 | 8 | 1402 | |||
| XM_011517448.2 | XP_011515750.1 | 7 | 1273 | |||
| XM_011517449.2 | XP_011515751.1 | 8 | 1402 | |||
| XM_017013019.1 | XP_016868508.1 | 7 | 1273 | |||
| GenBank mRNA | AB056652.1 | - | 7 | 2398 | ||
| AK094191.1 | - | 2 | 231 | |||
| BC041961.1 | - | 8 | 1430 | 5302156 | ||

C8orf34
Genomic Map