siRNA Details
| Gene Symbol: RBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052960.2 | NP_443192.1 | 2 | 299 | ||
| GenBank mRNA | AF399927.1 | - | 2 | 280 | ||
| AK312027.1 | - | 2 | 267 | |||
| AY145438.1 | - | 2 | 266 | |||
| BC033883.1 | - | 3 | 512 | |||
| BC063013.1 | - | 2 | 299 | 5224517 | ||

RBP7
Genomic Map