siRNA Details
| Gene Symbol: FAM129A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052966.3 | NP_443198.1 | 2 | 304 | ||
| XM_011509140.2 | XP_011507442.1 | 3 | 1179 | |||
| XM_011509141.2 | XP_011507443.1 | 3 | 1179 | |||
| GenBank mRNA | AB050477.1 | - | 2 | 230 | ||
| AF288391.1 | - | 2 | 268 | |||
| AK022527.1 | - | 2 | 142 | |||
| AK310557.1 | - | 2 | 171 | |||
| AK311019.1 | - | 2 | 171 | |||
| BC030531.1 | - | 2 | 160 | 5212338 | ||
| DC346827.1 | - | 2 | 304 | |||

FAM129A
Genomic Map