siRNA Details
| Gene Symbol: PAX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006193.2 | NP_006184.2 | 9 | 1261 | ||
| XM_011516276.2 | XP_011514578.1 | 12 | 1535 | |||
| GenBank mRNA | AF043978.1 | - | 9 | 1261 | ||
| BC107150.1 | - | 8 | 1510 | 40032102 | ||

PAX4
Genomic Map