siRNA Details
| Gene Symbol: ODF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286136.1 | NP_001273065.1 | 3, 4 | 612 | ||
| NM_053280.4 | NP_444510.2 | 3, 4 | 612 | |||
| GenBank mRNA | AB067774.1 | - | 3, 4 | 719 | ||
| AL133658.1 | - | 3 | 1001 | |||
| AY713299.1 | - | 3, 4 | 587 | |||
| BC126222.1 | - | 3, 4 | 420 | 8991938 | ||
| BC126248.1 | - | 3, 4 | 420 | 8991964 | ||
| BC144022.1 | - | 3, 4 | 420 | 9052542 | ||

ODF3
Genomic Map