siRNA Details
| Gene Symbol: ZSCAN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199479.1 | NP_001186408.1 | 5 | 958 | ||
| NM_001199480.1 | NP_001186409.1 | 4 | 769 | |||
| NM_006299.4 | NP_006290.1 | 4 | 805 | |||
| XM_011514875.2 | XP_011513177.1 | 5 | 1453 | |||
| XM_011514876.2 | XP_011513178.1 | 5 | 1434 | |||
| XM_011514878.2 | XP_011513180.1 | 4 | 1277 | |||
| XM_017011267.1 | XP_016866756.1 | 4 | 1256 | |||
| GenBank mRNA | AK226035.1 | - | 5 | 914 | ||
| AK304013.1 | - | 5 | 934 | |||
| AY550973.1 | - | 3 | 683 | |||
| BC010374.1 | - | 4 | 765 | 4132536 | ||
| U62392.1 | - | 4 | 717 | |||

ZSCAN9
Genomic Map