siRNA Details
| Gene Symbol: SLC30A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006345.3 | NP_006336.3 | 18 | 2974 | ||
| GenBank mRNA | AF006621.2 | - | 18 | 2948 | ||
| BC000240.1 | - | 18 | 2936 | |||
| BC016949.1 | - | 18 | 2953 | 3850521 | ||
| BC022981.1 | - | 14 | 2365 | |||

SLC30A9
Genomic Map