siRNA Details
| Gene Symbol: EPB41L4A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015370.2 | 1 | 655 | |||
| GenBank mRNA | AB048207.1 | - | 1 | 655 | ||
| BC101844.1 | - | 1 | 652 | |||
| BC112077.1 | - | 1 | 652 | |||
| DA266578.1 | - | 1 | 24 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPB41L4A-AS1| Gene Symbol: EPB41L4A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015370.2 | 1 | 655 | |||
| GenBank mRNA | AB048207.1 | - | 1 | 655 | ||
| BC101844.1 | - | 1 | 652 | |||
| BC112077.1 | - | 1 | 652 | |||
| DA266578.1 | - | 1 | 24 | |||