siRNA Details
| Gene Symbol: GALNT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319051.1 | NP_001305980.1 | 1 | 1209 | ||
| NM_054110.4 | NP_473451.3 | 1 | 854 | |||
| XM_005264852.4 | XP_005264909.1 | 1 | 386 | |||
| XM_017005682.1 | XP_016861171.1 | 1 | 386 | |||
| XM_017005683.1 | XP_016861172.1 | 1 | 386 | |||
| XM_017005684.1 | XP_016861173.1 | 1 | 386 | |||
| GenBank mRNA | AB078149.1 | - | 1 | 379 | ||
| AK225673.1 | - | 1 | 879 | |||
| AK312425.1 | - | 1 | 855 | |||
| AL831925.1 | - | 1 | 882 | |||
| AL832575.2 | - | 1 | 359 | |||
| AL832736.1 | - | 1 | 824 | |||
| AY035399.1 | - | 1 | 662 | |||
| AY358443.1 | - | 1 | 840 | |||
| BC014789.1 | - | 1 | 804 | 4212702 | ||

GALNT15
Genomic Map