siRNA Details
| Gene Symbol: KRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057088.2 | NP_476429.2 | 2 | 782 | ||
| GenBank mRNA | AJ628418.1 | - | 2 | 671 | ||
| AK314987.1 | - | 2 | 791 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT3| Gene Symbol: KRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057088.2 | NP_476429.2 | 2 | 782 | ||
| GenBank mRNA | AJ628418.1 | - | 2 | 671 | ||
| AK314987.1 | - | 2 | 791 | |||