siRNA Details
| Gene Symbol: DIRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052952.2 | NP_443184.1 | 2 | 1081 | ||
| GenBank mRNA | AY039011.1 | - | 2 | 1082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIRC1| Gene Symbol: DIRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052952.2 | NP_443184.1 | 2 | 1081 | ||
| GenBank mRNA | AY039011.1 | - | 2 | 1082 | ||