siRNA Details
| Gene Symbol: CYYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320768.1 | NP_001307697.1 | 4 | 2396 | ||
| NM_052954.3 | NP_443186.2 | 4 | 2393 | |||
| NR_135472.1 | 5 | 2563 | ||||
| GenBank mRNA | AF401639.1 | - | 4 | 1998 | ||
| AL833200.1 | - | 4 | 2204 | |||
| AY061853.1 | - | 4 | 1938 | |||

CYYR1
Genomic Map