siRNA Details
Gene Symbol: EPB41L4A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015370.2 | 3 | 929 | |||
GenBank mRNA | AB002437.1 | - | 3 | 561 | ||
AB048207.1 | - | 3 | 929 | |||
DA266578.1 | - | 3 | 297 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: EPB41L4A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015370.2 | 3 | 929 | |||
GenBank mRNA | AB002437.1 | - | 3 | 561 | ||
AB048207.1 | - | 3 | 929 | |||
DA266578.1 | - | 3 | 297 |