siRNA Details
| Gene Symbol: RBM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053043.2 | NP_444271.2 | 14 | 3013 | ||
| XM_005249531.1 | XP_005249588.1 | 15 | 3178 | |||
| XM_005249532.1 | XP_005249589.1 | 15 | 3178 | |||
| XM_005249533.1 | XP_005249590.1 | 15 | 3148 | |||
| XM_005249534.1 | XP_005249591.1 | 14 | 3058 | |||
| XM_005249535.1 | XP_005249592.1 | 14 | 3028 | |||
| XM_011515840.1 | XP_011514142.1 | 15 | 3178 | |||
| GenBank mRNA | AK301179.1 | - | 9 | 1960 | ||
| AL137724.1 | - | 12 | 3908 | |||

RBM33
Genomic Map