siRNA Details
| Gene Symbol: HTRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053044.4 | NP_444272.1 | 8 | 1355 | ||
| GenBank mRNA | AY040094.2 | - | 8 | 1372 | ||
| AY280665.1 | - | 8 | 1343 | |||
| BC034390.1 | - | 8 | 1355 | 5180585 | ||
| BC035717.1 | - | 8 | 1355 | 5588608 | ||

HTRA3
Genomic Map