siRNA Details
| Gene Symbol: TEKT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053285.1 | NP_444515.1 | 3 | 343 | ||
| XM_011524027.2 | XP_011522329.1 | 3 | 339 | |||
| GenBank mRNA | AF357879.1 | - | 3 | 292 | ||
| AK292818.1 | - | 3 | 308 | |||
| BC014599.2 | - | 3 | 343 | 4824860 | ||

TEKT1
Genomic Map