siRNA Details
| Gene Symbol: MGAT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014275.4 | NP_055090.1 | 10, 11 | 1526 | ||
| NM_054013.3 | NP_463459.1 | 9, 10 | 1966 | |||
| XM_017008998.1 | XP_016864487.1 | 10, 11 | 1352 | |||
| GenBank mRNA | AB000624.1 | - | 10, 11 | 1199 | ||
| AK023137.1 | - | 9, 10 | 1966 | |||
| AL162067.1 | - | 7, 8 | 630 | |||
| AY358984.1 | - | 10, 11 | 1207 | |||
| BC009464.1 | - | 8, 9 | 853 | |||
| BC051835.1 | - | 10, 11 | 1402 | 6196058 | ||

MGAT4B
Genomic Map