siRNA Details
| Gene Symbol: TRIM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003818.2 | NP_001003818.1 | 8 | 3178 | ||
| NM_001198644.1 | NP_001185573.1 | 7 | 2688 | |||
| NM_001198645.1 | NP_001185574.1 | 6 | 2592 | |||
| NM_058166.4 | NP_477514.1 | 8 | 2979 | |||
| GenBank mRNA | AF220030.1 | - | 8 | 2967 | ||
| AK023210.1 | - | 5 | 2230 | |||
| BC047564.1 | - | 8 | 3158 | 4825118 | ||
| BC065575.1 | - | 8 | 2704 | 4609084 | ||

TRIM6
Genomic Map