siRNA Details
| Gene Symbol: BORCS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300742.1 | NP_001287671.1 | 4 | 913 | ||
| NM_058169.4 | NP_477517.1 | 4 | 1304 | |||
| XM_011520551.2 | XP_011518853.1 | 4 | 947 | |||
| XM_011520552.2 | XP_011518854.1 | 3 | 1134 | |||
| GenBank mRNA | AK075028.1 | - | 4 | 1261 | ||
| BC013668.1 | - | 4 | 1187 | 4154039 | ||

BORCS5
Genomic Map