siRNA Details
| Gene Symbol: SMIM11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058182.4 | NP_478062.1 | 3 | 313 | ||
| XM_017028397.1 | XP_016883886.1 | 3 | 272 | |||
| XM_017028398.1 | XP_016883887.1 | 3 | 272 | |||
| XM_017028399.1 | XP_016883888.1 | 3 | 821 | |||
| GenBank mRNA | AK092693.1 | - | 3 | 248 | ||
| AY033902.1 | - | 2 | 195 | |||
| AY081144.1 | - | 3 | 263 | |||
| BC015596.2 | - | 3 | 244 | 4647871 | ||
| Gene Symbol: SMIM11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001313692.1 | NP_001300621.1 | 3 | 283 | ||
| XM_017028203.1 | XP_016883692.1 | 3 | 272 | |||
| XM_017028204.1 | XP_016883693.1 | 3 | 272 | |||
| XM_017028205.1 | XP_016883694.1 | 3 | 821 | |||
| GenBank mRNA | AW901268.1 | - | 3 | 250 | ||
| BP194116.1 | - | 3 | 283 | |||

SMIM11A
Genomic Map