siRNA Details
| Gene Symbol: MYL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021019.4 | NP_066299.2 | 3 | 216 | ||
| NM_079423.3 | NP_524147.2 | 3 | 216 | |||
| GenBank mRNA | AB046613.1 | - | 3 | 115 | ||
| AK301198.1 | - | 2 | 394 | |||
| BC006781.2 | - | 1 | 46 | |||
| BC017455.1 | - | 3 | 103 | 3538373 | ||
| BC071661.1 | - | 3 | 99 | 5561575 | ||
| BC093066.1 | - | 3 | 112 | 30722094 | ||
| BG283793.1 | - | 3 | 216 | |||
| CN401857.1 | - | 3 | 100 | |||
| CR457014.1 | - | 2 | 75 | |||
| M22918.1 | - | 3 | 106 | |||
| M22920.1 | - | 2 | 84 | |||
| M31212.1 | - | 3 | 115 | |||
| U02629.1 | - | 3 | 113 | |||

MYL6
Genomic Map