siRNA Details
| Gene Symbol: SELM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080430.2 | NP_536355.1 | 2, 3 | 230 | ||
| GenBank mRNA | AY043487.1 | - | 2, 3 | 259 | ||
| BC013421.1 | - | 2, 3 | 176 | 3890282 | ||
| BC030236.1 | - | 2, 3 | 187 | 4184342 | ||
| BC068004.1 | - | 2, 3 | 220 | 6058758 | ||

SELM
Genomic Map