siRNA Details
| Gene Symbol: CCM2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080625.3 | NP_542192.2 | 6 | 990 | ||
| XM_006723706.3 | XP_006723769.1 | 6 | 1053 | |||
| XM_011528566.1 | XP_011526868.1 | 6 | 1053 | |||
| XM_011528567.2 | XP_011526869.1 | 6 | 1053 | |||
| XM_011528568.2 | XP_011526870.1 | 7 | 1179 | |||
| XM_011528569.1 | XP_011526871.1 | 6 | 1053 | |||
| GenBank mRNA | AK057090.1 | - | 7 | 770 | ||
| AK125588.1 | - | 1 | 329 | |||
| BC030254.1 | - | 6 | 982 | 5182496 | ||
| BC032455.1 | - | 2 | 180 | |||

CCM2L
Genomic Map