siRNA Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080626.5 | NP_542193.3 | 2 | 375 | ||
| XM_011537940.2 | XP_011536242.1 | 2 | 363 | |||
| GenBank mRNA | AF283671.2 | - | 2 | 114 | ||
| AF284094.1 | - | 2 | 319 | |||
| BC034525.1 | - | 2 | 352 | 4778946 | ||
| CA488612.1 | - | 386 | ||||

BRI3BP
Genomic Map