siRNA Details
| Gene Symbol: ABCA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080282.3 | NP_525021.3 | 7 | 1217 | ||
| GenBank mRNA | AK310858.1 | - | 5 | 643 | ||
| AL832004.2 | - | 6 | 725 | |||
| AY028900.1 | - | 7 | 1217 | |||
| AY247065.1 | - | 7 | 1248 | |||
| BC136460.1 | - | 4 | 422 | |||
| BC144124.1 | - | 4 | 422 | |||

ABCA10
Genomic Map